N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051493-500MG 051493-2.5G
101895-864EA 364.62 USD
101895-864 101895-866
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinamine dihydrochloride
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂O
MDL Number: MFCD13561562
CAS Number: 1219957-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR