N-Benzyl-N-ethyl-3-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051492-500MG 051492-2.5G
101895-860EA 364.62 USD
101895-860 101895-862
N-Benzyl-N-ethyl-3-piperidinamine dihydrochloride
N-Benzyl-N-ethyl-3-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₄Cl₂N₂
MDL Number: MFCD13561561
CAS Number: 1220019-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR