1-(3-Piperidinyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Supplier: Matrix Scientific

051491-500MG 051491-2.5G
101895-856EA 364.62 USD
101895-856 101895-858
1-(3-Piperidinyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
1-(3-Piperidinyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂
MDL Number: MFCD13561560
CAS Number: 1219979-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR