1-(3-Piperidinyl)indoline dihydrochloride
Supplier: Matrix Scientific
051490-500MG
051490-2.5G
101895-852EA
364.62
USD
101895-852
101895-854
1-(3-Piperidinyl)indoline dihydrochloride
1-(3-Piperidinyl)indoline dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀Cl₂N₂ MW: 275.22 g/mol |
MDL Number:
MFCD13561559 CAS Number: 1219957-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...