2-[4-(3-Piperidinyl)-1-piperazinyl]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051488-500MG 051488-2.5G
101895-844EA 364.62 USD
101895-844 101895-846
2-[4-(3-Piperidinyl)-1-piperazinyl]-1-ethanol dihydrochloride
2-[4-(3-Piperidinyl)-1-piperazinyl]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅Cl₂N₃O
MDL Number: MFCD13561557
CAS Number: 1219979-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR