N-Benzyl-N-methyl-3-piperidinamine dihydrochloride
Supplier: Matrix Scientific
051486-500MG
051486-2.5G
101895-836EA
461.54
USD
101895-836
101895-838
N-Benzyl-N-methyl-3-piperidinamine dihydrochloride
N-Benzyl-N-methyl-3-piperidinamine dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₂Cl₂N₂ |
MDL Number:
MFCD13561555 CAS Number: 1220037-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...