3-(2-Ethyl-1-piperidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051485-500MG 051485-2.5G
101895-832EA 364.62 USD
101895-832 101895-834
3-(2-Ethyl-1-piperidinyl)piperidine dihydrochloride
3-(2-Ethyl-1-piperidinyl)piperidine dihydrochloride
Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561554
CAS Number: 1219979-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR