N-Cyclohexyl-N-methyl-3-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051484-500MG 051484-2.5G
101895-828EA 364.62 USD
101895-828 101895-830
N-Cyclohexyl-N-methyl-3-piperidinamine dihydrochloride
N-Cyclohexyl-N-methyl-3-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561553
CAS Number: 1220017-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR