N,N-Dipropyl-3-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051483-500MG 051483-2.5G
101895-824EA 364.62 USD
101895-824 101895-826
N,N-Dipropyl-3-piperidinamine dihydrochloride
N,N-Dipropyl-3-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₆Cl₂N₂
MDL Number: MFCD13561552
CAS Number: 1220017-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR