1-(3-Piperidinyl)-4-piperidinol dihydrochloride
Supplier: Matrix Scientific
051482-500MG
051482-2.5G
101895-820EA
364.62
USD
101895-820
101895-822
1-(3-Piperidinyl)-4-piperidinol dihydrochloride
1-(3-Piperidinyl)-4-piperidinol dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₂Cl₂N₂O MW: 257.21 g/mol |
MDL Number:
MFCD13561551 CAS Number: 1220019-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...