1-(3-Piperidinyl)azepane dihydrochloride

Supplier: Matrix Scientific

051480-500MG 051480-2.5G
101895-812EA 364.62 USD
101895-812 101895-814
1-(3-Piperidinyl)azepane dihydrochloride
1-(3-Piperidinyl)azepane dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MW: 255.23 g/mol
MDL Number: MFCD13561549
CAS Number: 1219957-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR