1-Benzyl-4-(3-piperidinyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051479-500MG 051479-2.5G
101895-808EA 364.62 USD
101895-808 101895-810
1-Benzyl-4-(3-piperidinyl)piperazine dihydrochloride
1-Benzyl-4-(3-piperidinyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₇Cl₂N₃
MDL Number: MFCD13561548
CAS Number: 1219979-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR