3-(3-Methyl-1-piperidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051476-500MG 051476-2.5G
101895-796EA 364.62 USD
101895-796 101895-798
3-(3-Methyl-1-piperidinyl)piperidine dihydrochloride
3-(3-Methyl-1-piperidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561545
CAS Number: 1220019-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR