3-(2-Methyl-1-piperidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051475-500MG 051475-2.5G
101895-792EA 364.62 USD
101895-792 101895-794
3-(2-Methyl-1-piperidinyl)piperidine dihydrochloride
3-(2-Methyl-1-piperidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MW: 255.23 g/mol
MDL Number: MFCD13561544
CAS Number: 1220030-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR