2-[Ethyl(3-piperidinyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051473-500MG 051473-2.5G
101895-784EA 364.62 USD
101895-784 101895-786
2-[Ethyl(3-piperidinyl)amino]-1-ethanol dihydrochloride
2-[Ethyl(3-piperidinyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂O
MDL Number: MFCD13561542
CAS Number: 1220017-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR