N-Butyl-N-methyl-3-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051472-500MG 051472-2.5G
101895-780EA 364.62 USD
101895-780 101895-782
N-Butyl-N-methyl-3-piperidinamine dihydrochloride
N-Butyl-N-methyl-3-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂
MDL Number: MFCD13561541
CAS Number: 1220019-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR