1-(3-Piperidinyl)-3-pyrrolidinol dihydrochloride

Supplier: Matrix Scientific

051471-500MG 051471-2.5G
101895-776EA 364.62 USD
101895-776 101895-778
1-(3-Piperidinyl)-3-pyrrolidinol dihydrochloride
1-(3-Piperidinyl)-3-pyrrolidinol dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂O
MDL Number: MFCD13561540
CAS Number: 1219957-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR