4-(3-Piperidinyl)morpholine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 4-(Piperidin-3-yl)morpholine dihydrochloride

051470-500MG 051470-2.5G
101895-772EA 384.62 USD
101895-772 101895-774
4-(3-Piperidinyl)morpholine dihydrochloride
4-(3-Piperidinyl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂O
MDL Number: MFCD13561539
CAS Number: 1124199-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR