3-(1-Piperidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051469-500MG 051469-2.5G
101895-768EA 166.15 USD
101895-768 101895-770
3-(1-Piperidinyl)piperidine dihydrochloride
3-(1-Piperidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂
MW: 241.21 g/mol
MDL Number: MFCD13561538
CAS Number: 813425-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR