N,N-Diethyl-3-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051467-500MG 051467-2.5G
101895-760EA 364.62 USD
101895-760 101895-762
N,N-Diethyl-3-piperidinamine dihydrochloride
N,N-Diethyl-3-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂
MDL Number: MFCD13561536
CAS Number: 1220034-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR