N-Methyl(4-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride

Supplier: Matrix Scientific

051466-500MG 051466-2.5G
101895-756EA 364.62 USD
101895-756 101895-758
N-Methyl(4-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride
N-Methyl(4-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂O
MW: 299.29 g/mol
MDL Number: MFCD13561535
CAS Number: 1220030-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR