N-Benzyl-N-(4-piperidinylmethyl)-1-ethanamine dihydrochloride
Supplier: Matrix Scientific
051465-500MG
051465-2.5G
101895-752EA
364.62
USD
101895-752
101895-754
N-Benzyl-N-(4-piperidinylmethyl)-1-ethanamine dihydrochloride
N-Benzyl-N-(4-piperidinylmethyl)-1-ethanamine dihydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₆Cl₂N₂ MW: 305.29 g/mol |
MDL Number:
MFCD13561534 CAS Number: 1220017-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...