1-(4-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Supplier: Matrix Scientific

051464-500MG 051464-2.5G
101895-748EA 292.31 USD
101895-748 101895-750
1-(4-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
1-(4-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄Cl₂N₂
MDL Number: MFCD13561533
CAS Number: 1220037-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR