1-(4-Piperidinylmethyl)indoline dihydrochloride

Supplier: Matrix Scientific

051463-500MG 051463-2.5G
101895-744EA 364.62 USD
101895-744 101895-746
1-(4-Piperidinylmethyl)indoline dihydrochloride
1-(4-Piperidinylmethyl)indoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂
MW: 289.25 g/mol
MDL Number: MFCD13561532
CAS Number: 1220038-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR