2-(4-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
Supplier: Matrix Scientific
051462-500MG
051462-2.5G
101895-740EA
384.62
USD
101895-740
101895-742
2-(4-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
2-(4-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄Cl₂N₂ |
MDL Number:
MFCD13561531 CAS Number: 120847-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...