N-Ethyl-N-(4-piperidinylmethyl)cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051460-500MG 051460-2.5G
101895-732EA 364.62 USD
101895-732 101895-734
N-Ethyl-N-(4-piperidinylmethyl)cyclohexanamine dihydrochloride
N-Ethyl-N-(4-piperidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₃₀Cl₂N₂
MDL Number: MFCD13561529
CAS Number: 1220016-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR