N-Methyl(phenyl)-N-(4-piperidinylmethyl)methanamine dihydrochloride
Supplier: Matrix Scientific
051459-500MG
051459-2.5G
101895-728EA
364.62
USD
101895-728
101895-730
N-Methyl(phenyl)-N-(4-piperidinylmethyl)methanamine dihydrochloride
N-Methyl(phenyl)-N-(4-piperidinylmethyl)methanamine dihydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₄Cl₂N₂ MW: 291.27 g/mol |
MDL Number:
MFCD13561528 CAS Number: 1211485-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...