2-Ethyl-1-(4-piperidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051458-500MG 051458-2.5G
101895-724EA 364.62 USD
101895-724 101895-726
2-Ethyl-1-(4-piperidinylmethyl)piperidine dihydrochloride
2-Ethyl-1-(4-piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561527
CAS Number: 1220034-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR