N-Methyl-N-(4-piperidinylmethyl)cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051457-500MG 051457-2.5G
101895-720EA 364.62 USD
101895-720 101895-722
N-Methyl-N-(4-piperidinylmethyl)cyclohexanamine dihydrochloride
N-Methyl-N-(4-piperidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561526
CAS Number: 1220017-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR