1-(4-Piperidinylmethyl)-4-piperidinol dihydrochloride

Supplier: Matrix Scientific

051455-500MG 051455-2.5G
101895-712EA 364.62 USD
101895-712 101895-714
1-(4-Piperidinylmethyl)-4-piperidinol dihydrochloride
1-(4-Piperidinylmethyl)-4-piperidinol dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂O
MW: 271.23 g/mol
MDL Number: MFCD13561524
CAS Number: 1220037-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR