1-(4-Piperidinylmethyl)azepane dihydrochloride

Supplier: Matrix Scientific

051453-500MG 051453-2.5G
101895-704EA 364.62 USD
101895-704 101895-706
1-(4-Piperidinylmethyl)azepane dihydrochloride
1-(4-Piperidinylmethyl)azepane dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13469304
CAS Number: 1211466-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR