1-Benzyl-4-(4-piperidinylmethyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051452-500MG 051452-2.5G
101895-700EA 364.62 USD
101895-700 101895-702
1-Benzyl-4-(4-piperidinylmethyl)piperazine dihydrochloride
1-Benzyl-4-(4-piperidinylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₇H₂₉Cl₂N₃
MDL Number: MFCD13561522
CAS Number: 1219979-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR