1-Methyl-4-(4-piperidinylmethyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051451-500MG 051451-2.5G
101895-696EA 384.62 USD
101895-696 101895-698
1-Methyl-4-(4-piperidinylmethyl)piperazine dihydrochloride
1-Methyl-4-(4-piperidinylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅Cl₂N₃
MDL Number: MFCD13561521
CAS Number: 1219957-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR