3-Methyl-1-(4-piperidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051449-500MG 051449-2.5G
101895-688EA 166.15 USD
101895-688 101895-690
3-Methyl-1-(4-piperidinylmethyl)piperidine dihydrochloride
3-Methyl-1-(4-piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561519
CAS Number: 1172842-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR