2-Methyl-1-(4-piperidinylmethyl)piperidine dihydrochloride
Supplier: Matrix Scientific
051448-500MG
051448-2.5G
101895-684EA
364.62
USD
101895-684
101895-686
2-Methyl-1-(4-piperidinylmethyl)piperidine dihydrochloride
2-Methyl-1-(4-piperidinylmethyl)piperidine dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆Cl₂N₂ MW: 269.26 g/mol |
MDL Number:
MFCD13561518 CAS Number: 1211495-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...