N-Methyl-N-(4-piperidinylmethyl)-1-butanamine dihydrochloride

Supplier: Matrix Scientific

051445-500MG 051445-2.5G
101895-672EA 364.62 USD
101895-672 101895-674
N-Methyl-N-(4-piperidinylmethyl)-1-butanamine dihydrochloride
N-Methyl-N-(4-piperidinylmethyl)-1-butanamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₆Cl₂N₂
MDL Number: MFCD13561515
CAS Number: 1220030-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR