1-(4-Piperidinylmethyl)-3-pyrrolidinol dihydrochloride
Supplier: Matrix Scientific
051444-500MG
051444-2.5G
101895-668EA
364.62
USD
101895-668
101895-670
1-(4-Piperidinylmethyl)-3-pyrrolidinol dihydrochloride
1-(4-Piperidinylmethyl)-3-pyrrolidinol dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₂Cl₂N₂O MW: 257.21 g/mol |
MDL Number:
MFCD13561514 CAS Number: 1220030-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...