4-(4-Piperidinylmethyl)morpholine dihydrochloride

Supplier: Matrix Scientific

051443-500MG 051443-2.5G
101895-664EA 461.54 USD
101895-664 101895-666
4-(4-Piperidinylmethyl)morpholine dihydrochloride
4-(4-Piperidinylmethyl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂O
MDL Number: MFCD13561513
CAS Number: 81310-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR