1-(4-Piperidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051442-500MG 051442-2.5G
101895-660EA 364.62 USD
101895-660 101895-662
1-(4-Piperidinylmethyl)piperidine dihydrochloride
1-(4-Piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD12407022
CAS Number: 32832-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR