2-[Methyl-(4-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051441-500MG 051441-2.5G
101895-656EA 364.62 USD
101895-656 101895-658
2-[Methyl-(4-piperidinylmethyl)amino]-1-ethanol dihydrochloride
2-[Methyl-(4-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂O
MDL Number: MFCD13561512
CAS Number: 1220030-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR