N-Ethyl-N-(4-piperidinylmethyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051440-500MG 051440-2.5G
101895-652EA 364.62 USD
101895-652 101895-654
N-Ethyl-N-(4-piperidinylmethyl)-1-ethanamine dihydrochloride
N-Ethyl-N-(4-piperidinylmethyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂
MDL Number: MFCD13561511
CAS Number: 614746-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR