4-(1-Pyrrolidinylmethyl)piperidine dihydrochloride
Supplier: Matrix Scientific
Synonyms:
4-(Pyrrolidin-1-ylmethyl)piperidine dihydrochloride
051439-500MG
051439-2.5G
101895-648EA
364.62
USD
101895-648
101895-650
4-(1-Pyrrolidinylmethyl)piperidine dihydrochloride
4-(1-Pyrrolidinylmethyl)piperidine dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₂Cl₂N₂ MW: 241.21 g/mol |
MDL Number:
MFCD12407023 CAS Number: 780756-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...