N,N-Dimethyl(4-piperidinyl)methanamine dihydrochloride

Supplier: Matrix Scientific

051438-500MG 051438-2.5G
101895-644EA 364.62 USD
101895-644 101895-646
N,N-Dimethyl(4-piperidinyl)methanamine dihydrochloride
N,N-Dimethyl(4-piperidinyl)methanamine dihydrochloride

Hazard: Irritant

Formula: C₈H₂₀Cl₂N₂
MDL Number: MFCD13561510
CAS Number: 172281-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR