N-Methyl(3-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride
Supplier: Matrix Scientific
051437-500MG
051437-2.5G
101895-640EA
364.62
USD
101895-640
101895-642
N-Methyl(3-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride
N-Methyl(3-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₈Cl₂N₂O MW: 299.29 g/mol |
MDL Number:
MFCD13561509 CAS Number: 1220038-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...