2-(3-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Supplier: Matrix Scientific

051433-500MG 051433-2.5G
101895-624EA 364.62 USD
101895-624 101895-626
2-(3-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
2-(3-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄Cl₂N₂
MDL Number: MFCD13561505
CAS Number: 1220019-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR