2-[4-(3-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051432-500MG 051432-2.5G
101895-620EA 364.62 USD
101895-620 101895-622
2-[4-(3-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride
2-[4-(3-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₇Cl₂N₃O
MDL Number: MFCD13561504
CAS Number: 1220029-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR