N-Ethyl-N-(3-piperidinylmethyl)cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051431-500MG 051431-2.5G
101895-616EA 364.62 USD
101895-616 101895-618
N-Ethyl-N-(3-piperidinylmethyl)cyclohexanamine dihydrochloride
N-Ethyl-N-(3-piperidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₃₀Cl₂N₂
MDL Number: MFCD13561503
CAS Number: 1220034-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR