N-Methyl-N-(3-piperidinylmethyl)cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051428-500MG 051428-2.5G
101895-604EA 364.62 USD
101895-604 101895-606
N-Methyl-N-(3-piperidinylmethyl)cyclohexanamine dihydrochloride
N-Methyl-N-(3-piperidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561500
CAS Number: 1220016-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR