N-(3-Piperidinylmethyl)-N-propyl-1-propanamine dihydrochloride

Supplier: Matrix Scientific

051427-500MG 051427-2.5G
101895-600EA 364.62 USD
101895-600 101895-602
N-(3-Piperidinylmethyl)-N-propyl-1-propanamine dihydrochloride
N-(3-Piperidinylmethyl)-N-propyl-1-propanamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₈Cl₂N₂
MW: 271.28 g/mol
MDL Number: MFCD13561499
CAS Number: 1219957-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR