1-(3-Piperidinylmethyl)-4-piperidinol dihydrochloride
Supplier: Matrix Scientific
051426-500MG
051426-2.5G
101895-596EA
364.62
USD
101895-596
101895-598
1-(3-Piperidinylmethyl)-4-piperidinol dihydrochloride
1-(3-Piperidinylmethyl)-4-piperidinol dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₄Cl₂N₂O MW: 271.23 g/mol |
MDL Number:
MFCD13561498 CAS Number: 1220029-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...